CAS RN 110878-71-2
IUPAC Name:
3-(piperidin-4-yl)phenol
Synonyms:
3-(piperidin-4-yl)phenol
Molecular Weight (g/mol):
177.25
Molecular Formula:
C11H15NO
InChi Key:
FINLIMGQGJZNRN-UHFFFAOYSA-N
SMILES:
OC1=CC=CC(=C1)C1CCNCC1
1
–
1
of
1
results
1
4-(3-Hydroxyphenyl)piperidine 98.0+%, TCI America™
CAS: 110878-71-2 Molecular Formula: C11H15NO Molecular Weight (g/mol): 177.247 MDL Number: MFCD02093474 InChI Key: FINLIMGQGJZNRN-UHFFFAOYSA-N Synonym: 3-(4-Piperidyl)phenol PubChem CID: 10583453 IUPAC Name: 3-piperidin-4-ylphenol SMILES: C1CNCCC1C2=CC(=CC=C2)O