accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFFFFFFFFFFFFFFF

CAS RN 1109-15-5

IUPAC Name: tris(2,3,4,5,6-pentafluorophenyl)borane
Synonyms: tris(pentafluorophenyl)borane
Molecular Weight (g/mol): 511.98
Molecular Formula: C18BF15
InChi Key: OBAJXDYVZBHCGT-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(B(C2=C(F)C(F)=C(F)C(F)=C2F)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results