accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CONH2HCl

CAS RN 11099-03-9

IUPAC Name: 2-amino-N,N-dipropylacetamide hydrochloride
Synonyms: 2-amino-N,N-dipropylacetamide hydrochloride
Molecular Weight (g/mol): 194.7
Molecular Formula: C8H19ClN2O
InChi Key: GBPWTMZRCAESKB-UHFFFAOYSA-N
SMILES: Cl.CCCN(CCC)C(=O)CN
Color 
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
Grade 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)