accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3

CAS RN 111-06-8

IUPAC Name: butyl hexadecanoate
Synonyms: butyl palmitate
Molecular Weight (g/mol): 312.54
Molecular Formula: C20H40O2
InChi Key: GLYJVQDYLFAUFC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results