accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 111-16-0

IUPAC Name: heptanedioic acid
Synonyms: pimelic acid pilerate pileric acid
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: WLJVNTCWHIRURA-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCC(O)=O
Molecular Weight (g/mol) 
  • (12)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (5)
  • (7)
Quantity 
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (5)
  • (5)
Boiling Point 
  • (4)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (5)
  • (2)
  • (5)
  • (3)
  • (7)
  • (12)
  • (5)
  • (2)
  • (3)
  • (5)
  • (7)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (5)
  • (4)
  • (3)
  • (3)
  • (5)