accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-24-0

IUPAC Name: 1,5-dibromopentane
Synonyms: pentane, 1,5-dibromo-
Molecular Weight (g/mol): 229.94
Molecular Formula: C5H10Br2
InChi Key: IBODDUNKEPPBKW-UHFFFAOYSA-N
SMILES: BrCCCCCBr
Boiling Point 
  • (2)
  • (3)
  • (5)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (5)
  • (4)
  • (6)
  • (3)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)