accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-51-3

IUPAC Name: [4-(dimethylazaniumyl)butyl]dimethylazanium
Synonyms: [4-(dimethylammonio)butyl]dimethylazanium
Molecular Weight (g/mol): 146.28
Molecular Formula: C8H22N2
InChi Key: VEAZEPMQWHPHAG-UHFFFAOYSA-P
SMILES: C[NH+](C)CCCC[NH+](C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results