accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-60-4

IUPAC Name: 2-hydroxyethyl octadecanoate
Synonyms: glycol stearate lipal 400-S polyethylene monostearate #6000 glycol nonex 28 nonex 29 nonex 36 nonex 53 nonex 54 nonex 63 myrj solution
Molecular Weight (g/mol): 328.54
Molecular Formula: C20H40O3
InChi Key: RFVNOJDQRGSOEL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCCO
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results