accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3(E)

CAS RN 111-62-6

IUPAC Name: ethyl (9E)-octadec-9-enoate
Synonyms: ethyl oleate
Molecular Weight (g/mol): 310.52
Molecular Formula: C20H38O2
InChi Key: LVGKNOAMLMIIKO-VAWYXSNFSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)OCC
Molecular Weight (g/mol) 
  • (7)
  • (4)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (4)
  • (5)
Physical Form 
  • (8)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (4)
  • (4)
  • (4)
  • (4)
  • (6)
  • (3)
  • (7)
  • (4)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (5)
  • (8)
  • (3)
  • (4)
  • (3)