accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-67-1

IUPAC Name: (2Z)-oct-2-ene
Synonyms: (2Z)-oct-2-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: ILPBINAXDRFYPL-HYXAFXHYSA-N
SMILES: CCCCC\C=C/C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)