accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 111-71-7

IUPAC Name: heptanal
Synonyms: heptanal heptanal (natural) fema number 2541 heptanal, compd. with sulfurous acid monosodium heptaldehyde (substance) oenanthaldehyde enanthic aldehyde aldehyde C-7 oenanthic aldehyde oenanthol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: FXHGMKSSBGDXIY-UHFFFAOYSA-N
SMILES: CCCCCCC=O
Boiling Point 
  • (4)
  • (4)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (1)
  • (1)
  • (2)
  • (8)
  • (4)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (6)
  • (3)
  • (1)