accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNHN

CAS RN 111-94-4

IUPAC Name: 3-[(2-cyanoethyl)amino]propanenitrile
Synonyms: propanenitrile, 3,3'-iminobis-
Molecular Weight (g/mol): 123.16
Molecular Formula: C6H9N3
InChi Key: SBAJRGRUGUQKAF-UHFFFAOYSA-N
SMILES: N#CCCNCCC#N
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)