accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 11104-88-4

IUPAC Name: nickel(2+) molybdenum hydroxide oxidandiide phosphate
Synonyms: nickel(2+) molybdenum hydroxide oxidandiide phosphate
Molecular Weight (g/mol): 282.62
Molecular Formula: HMoNiO6P
InChi Key: KNQDGGOTQNRXGB-UHFFFAOYSA-J
SMILES: [OH-].[O--].[Ni++].[Mo].[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)