CAS RN 111051-56-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Ser Trt -OH, Advanced ChemTech
CAS: 111051-56-4 Molecular Formula: C37H31NO5 Molecular Weight (g/mol): 569.66