accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4H2ONH2OO

CAS RN 1111-78-0

IUPAC Name: ammonium hydrate carbamate
Synonyms: carbamic acid
Molecular Weight (g/mol): 96.09
Molecular Formula: CH8N2O3
InChi Key: SFWLALIEAXEULE-UHFFFAOYSA-N
SMILES: [NH4+].O.NC([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)