accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1111-92-8

IUPAC Name: dimethylphosphinoyl chloride
Synonyms: dimethylphosphinoyl chloride
Molecular Weight (g/mol): 112.49
Molecular Formula: C2H6ClOP
InChi Key: CVNMBKFJYRAHPO-UHFFFAOYSA-N
SMILES: CP(C)(Cl)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results