accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2(S)ClClOHO

CAS RN 111119-36-9

IUPAC Name: (2S)-2-amino-3-(2,4-dichlorophenyl)propanoic acid
Synonyms: (2S)-2-amino-3-(2,4-dichlorophenyl)propanoic acid
Molecular Weight (g/mol): 234.08
Molecular Formula: C9H9Cl2NO2
InChi Key: GWHQTNKPTXDNRM-QMMMGPOBSA-N
SMILES: N[C@@H](CC1=CC=C(Cl)C=C1Cl)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)