accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNSOONH2

CAS RN 111124-90-4

IUPAC Name: 1-methyl-1H-imidazole-4-sulfonamide
Synonyms: 1-methylimidazole-4-sulfonamide
Molecular Weight (g/mol): 161.18
Molecular Formula: C4H7N3O2S
InChi Key: QBJSSOBNVYDCDQ-UHFFFAOYSA-N
SMILES: CN1C=NC(=C1)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results