accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1111733-07-3

IUPAC Name: potassium trifluoro(isoquinolin-4-yl)boranuide
Synonyms: potassium trifluoro(isoquinolin-4-yl)boranuide
Molecular Weight (g/mol): 235.06
Molecular Formula: C9H6BF3KN
InChi Key: QEIOJGBGINOBGG-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=C2C=CC=CC2=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results