accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CH3CH2C

CAS RN 1112-54-5

IUPAC Name: ethenyltriethylsilane
Synonyms: ethenyltriethylsilane
Molecular Weight (g/mol): 142.32
Molecular Formula: C8H18Si
InChi Key: HBWGDHDXAMFADB-UHFFFAOYSA-N
SMILES: CC[Si](CC)(CC)C=C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results