accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2SnH2CH2CH2C

CAS RN 1112-56-7

IUPAC Name: tetraethenylstannane
Synonyms: tetraethenylstannane
Molecular Weight (g/mol): 226.89
Molecular Formula: C8H12Sn
InChi Key: MZIYQMVHASXABC-UHFFFAOYSA-N
SMILES: C=C[Sn](C=C)(C=C)C=C
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)