accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1112-66-9

CH2SiH2CH2CH2C

CAS RN 1112-66-9

IUPAC Name: tetrakis(prop-2-en-1-yl)silane
Synonyms: tetrakis(prop-2-en-1-yl)silane
Molecular Weight (g/mol): 192.38
Molecular Formula: C12H20Si
InChi Key: AKRQMTFHUVDMIL-UHFFFAOYSA-N
SMILES: C=CC[Si](CC=C)(CC=C)CC=C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)