accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH4NH4OWOOO

CAS RN 11120-25-5

IUPAC Name: diammonium dioxotungstenbis(olate)
Synonyms: ammonium paratungstate
Molecular Weight (g/mol): 283.91
Molecular Formula: H8N2O4W
InChi Key: IZYSFFFJIOTJEV-UHFFFAOYSA-P
SMILES: [NH4+].[NH4+].[O-][W]([O-])(=O)=O
Grade 
  • (1)
  • (2)
Solvent or Matrix 
  • (1)
Technique 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)