accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111247-62-2

IUPAC Name: 2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
Synonyms: 2-[4-(chloromethyl)piperidin-1-yl]pyrimidine
Molecular Weight (g/mol): 211.69
Molecular Formula: C10H14ClN3
InChi Key: XEOONRMTEFSPSW-UHFFFAOYSA-N
SMILES: ClCC1CCN(CC1)C1=NC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results