CAS RN 11128-94-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amberlite™ IRN-77, ion exchange resin, nuclear grade
CAS: 11128-94-2 Molecular Formula: Styrene-DVB MDL Number: MFCD00145822