accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111296-90-3

NHHN

CAS RN 111296-90-3

IUPAC Name: 5H,7H-indolo[2,3-b]carbazole
Synonyms: 5H,7H-indolo[2,3-b]carbazole
Molecular Weight (g/mol): 256.31
Molecular Formula: C18H12N2
InChi Key: UVHIWKCVGUMHNB-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2C2=CC3=C(NC4=C3C=CC=C4)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results