accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1113-38-8

IUPAC Name: diammonium oxalate
Synonyms: ammonium oxalate
Molecular Weight (g/mol): 124.1
Molecular Formula: C2H8N2O4
InChi Key: VBIXEXWLHSRNKB-UHFFFAOYSA-N
SMILES: [NH4+].[NH4+].[O-]C(=O)C([O-])=O
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results