accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111331-82-9

OOOHOHNCH3H3CCH3

CAS RN 111331-82-9

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}benzoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]benzoic acid
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: SAPAUOFSCLCQJB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC(=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)