accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHHN

CAS RN 111390-22-8

IUPAC Name: N-ethylpyrrolidin-3-amine
Synonyms: N-ethylpyrrolidin-3-amine
Molecular Weight (g/mol): 114.19
Molecular Formula: C6H14N2
InChi Key: OPCPWFHLFKAUEA-UHFFFAOYNA-N
SMILES: CCNC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results