accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OSCH3

CAS RN 1114-71-2

IUPAC Name: N-butyl-N-ethyl(propylsulfanyl)formamide
Synonyms: pebulate tillam
Molecular Weight (g/mol): 203.34
Molecular Formula: C10H21NOS
InChi Key: SGEJQUSYQTVSIU-UHFFFAOYSA-N
SMILES: CCCCN(CC)C(=O)SCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results