CAS RN 11141-16-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
2
Aroclor 1232 (High Level), SPEX CertiPrep™
CAS: 11141-16-5
3
Aroclor 1232, SPEX CertiPrep™
CAS: 11141-16-5