accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOOHOHNHCl

CAS RN 111469-81-9

IUPAC Name: 22-(cyclopropylmethyl)-14-oxa-11,22-diazaheptacyclo[13.9.1.0¹,¹³.0²,²¹.0⁴,¹².0⁵,¹⁰.0¹⁹,²⁵]pentacosa-4(12),5,7,9,15,17,19(25)-heptaene-2,16-diol hydrochloride
Synonyms: 22-(cyclopropylmethyl)-14-oxa-11,22-diazaheptacyclo[13.9.1.0¹,¹³.0²,²¹.0⁴,¹².0⁵,¹⁰.0¹⁹,²⁵]pentacosa-4(12),5,7,9,15,17,19(25)-heptaene-2,16-diol hydrochloride
Molecular Weight (g/mol): 450.96
Molecular Formula: C26H27ClN2O3
InChi Key: KNJKRQXCFJCQHC-UHFFFAOYNA-N
SMILES: Cl.OC1=C2OC3C4=C(CC5(O)C6CC(C=C1)=C2C35CCN6CC1CC1)C1=CC=CC=C1N4

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results