accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OO(S)H3CNH2HCl

CAS RN 1115-59-9

IUPAC Name: hydrogen ethyl 2-aminopropanoate chloride
Synonyms: hydrogen ethyl 2-aminopropanoate chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: JCXLZWMDXJFOOI-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(C)N
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)