accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1115084-83-7

IUPAC Name: 1-[(prop-2-yn-1-yloxy)methyl]pyrene
Synonyms: 1-[(prop-2-yn-1-yloxy)methyl]pyrene
Molecular Weight (g/mol): 270.33
Molecular Formula: C20H14O
InChi Key: PHLBHJYVJNAPAY-UHFFFAOYSA-N
SMILES: C#CCOCC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results