accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111544-93-5

IUPAC Name: 1,3-diethyl 2-(2,4-dichlorophenyl)propanedioate
Synonyms: 1,3-diethyl 2-(2,4-dichlorophenyl)propanedioate
Molecular Weight (g/mol): 305.15
Molecular Formula: C13H14Cl2O4
InChi Key: FIGCCTUCURBNIF-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C(=O)OCC)C1=CC=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results