accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1115639-92-3

IUPAC Name: 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 430.35
Molecular Formula: C30H27BO2
InChi Key: NZTBACVVLACPNL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results