accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1116-40-1

IUPAC Name: tris(2-methylpropyl)amine
Synonyms: tris(2-methylpropyl)amine
Molecular Weight (g/mol): 185.36
Molecular Formula: C12H27N
InChi Key: IIFFFBSAXDNJHX-UHFFFAOYSA-N
SMILES: CC(C)CN(CC(C)C)CC(C)C
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results