accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1116-77-4

CH3ONH3CCH3OCH3

CAS RN 1116-77-4

IUPAC Name: (4,4-diethoxybutyl)dimethylamine
Synonyms: (4,4-diethoxybutyl)dimethylamine
Molecular Weight (g/mol): 189.3
Molecular Formula: C10H23NO2
InChi Key: QKXMWBLNSPNBEY-UHFFFAOYSA-N
SMILES: CCOC(CCCN(C)C)OCC
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)