accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 1116499-73-0

IUPAC Name: 9-{3-[3'-(9H-carbazol-9-yl)-[1,1'-biphenyl]-3-yl]phenyl}-9H-carbazole
Synonyms: 9-{3-[3'-(carbazol-9-yl)-[1,1'-biphenyl]-3-yl]phenyl}carbazole
Molecular Weight (g/mol): 560.7
Molecular Formula: C42H28N2
InChi Key: KBYUHVUTGQEAEK-UHFFFAOYSA-N
SMILES: C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC(=C1)C1=CC(=CC=C1)C1=CC(=CC=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results