accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1117-86-8

IUPAC Name: octane-1,2-diol
Synonyms: 1,2-octanediol caprylyl glycol 1,2-octylene glycol
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: AEIJTFQOBWATKX-UHFFFAOYNA-N
SMILES: CCCCCCC(O)CO
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)