CAS RN 111755-37-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Microcystin-RR, Microcystis aeruginosa
Arg-Arg analog of Microcystin-LR that is less toxic. Inhibits protein phosphatase 2A (IC50 = 3.4 nM). Useful for comparative structure and function studies.