accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHNBr

CAS RN 111770-86-6

IUPAC Name: 5-bromo-2-ethynylpyridine
Synonyms: 5-bromo-2-ethynylpyridine
Molecular Weight (g/mol): 182.02
Molecular Formula: C7H4BrN
InChi Key: VGLAJERHBBEPQP-UHFFFAOYSA-N
SMILES: BrC1=CN=C(C=C1)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)