accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1118-02-1

IUPAC Name: isocyanatotrimethylsilane
Synonyms: isocyanatotrimethylsilane
Molecular Weight (g/mol): 115.21
Molecular Formula: C4H9NOSi
InChi Key: NIZHERJWXFHGGU-UHFFFAOYSA-N
SMILES: C[Si](C)(C)N=C=O
Boiling Point 
  • (3)
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (5)