accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2N(E)

CAS RN 1118-61-2

IUPAC Name: (2E)-3-aminobut-2-enenitrile
Synonyms: (2E)-3-aminobut-2-enenitrile
Molecular Weight (g/mol): 82.11
Molecular Formula: C4H6N2
InChi Key: DELJOESCKJGFML-DUXPYHPUSA-N
SMILES: C\C(N)=C/C#N
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)