accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1118-84-9

IUPAC Name: prop-2-en-1-yl 3-oxobutanoate
Synonyms: prop-2-en-1-yl 3-oxobutanoate
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: AXLMPTNTPOWPLT-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)OCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results