CAS RN 111821-49-9
CAS RN 111821-49-9
IUPAC Name:
(2R)-2-amino-3-[4-(dihydroxyboranyl)phenyl]propanoic acid
Synonyms:
(2R)-2-amino-3-[4-(dihydroxyboranyl)phenyl]propanoic acid
Molecular Weight (g/mol):
209.01
Molecular Formula:
C9H12BNO4
InChi Key:
NFIVJOSXJDORSP-MRVPVSSYSA-N
SMILES:
N[C@H](CC1=CC=C(C=C1)B(O)O)C(O)=O
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich ™-(+)-3,3'-Bis(3,5-Bis(Trifluoromethyl)Phenyl)-1,1'-Bi-2-Naphthol
Sigma Aldrich ™-(+)-3,3'-Bis(3,5-Bis(Trifluoromethyl)Phenyl)-1,1'-Bi-2-Naphthol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.