accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 1119-40-0

IUPAC Name: 1,5-dimethyl pentanedioate
Synonyms: methyl glutarate
Molecular Weight (g/mol): 160.17
Molecular Formula: C7H12O4
InChi Key: XTDYIOOONNVFMA-UHFFFAOYSA-N
SMILES: COC(=O)CCCC(=O)OC
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (5)