accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOCH2

CAS RN 1119-60-4

IUPAC Name: hept-6-enoic acid
Synonyms: 6-heptenoic acid 6-heptenoic acids epsilon-heptenoic acid hept-6-enoic acids epsilon-heptenoic acids
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: RWNJOXUVHRXHSD-UHFFFAOYSA-N
SMILES: OC(=O)CCCCC=C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)