accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1119-65-9

IUPAC Name: hept-2-yne
Synonyms: 2-heptyne
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: AMSFEMSYKQQCHL-UHFFFAOYSA-N
SMILES: CCCCC#CC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)