accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHOH

CAS RN 1119-86-4

IUPAC Name: decane-1,2-diol
Synonyms: decylene glycol
Molecular Weight (g/mol): 174.28
Molecular Formula: C10H22O2
InChi Key: YSRSBDQINUMTIF-UHFFFAOYNA-N
SMILES: CCCCCCCCC(O)CO
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)